Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM27 Mouse anti-Human, Clone: PCRP-TRIM27-1B3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP30741520UG
Description
TRIM27 Monoclonal specifically detects TRIM27 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
TRIM27 | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
Ret finger proteinRING finger protein 76, RFPtripartite motif protein TRIM27, RNF76RFP transforming protein, tripartite motif containing 27, tripartite motif-containing 27, Tripartite motif-containing protein 27, zinc finger protein RFP | |
Recombinant full-length human TRIM27 protein | |
20 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence, Peptide Array | |
PCRP-TRIM27-1B3 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
5987 | |
Store at 4°C. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title