Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM27 Mouse anti-Human, Clone: PCRP-TRIM27-1B3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30741510UG

 View more versions of this product

Catalog No. NB122303



Description

Description

TRIM27 Monoclonal antibody specifically detects TRIM27 in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Protein Array
Specifications

Specifications

TRIM27
PCRP-TRIM27-1B3
Unconjugated
10 mM PBS with 0.05% BSA
Ret finger proteinRING finger protein 76, RFPtripartite motif protein TRIM27, RNF76RFP transforming protein, tripartite motif containing 27, tripartite motif-containing 27, Tripartite motif-containing protein 27, zinc finger protein RFP
Recombinant full-length human TRIM27 protein
Protein A or G purified
RUO
Primary
5987
Flow Cytometry, Immunofluorescence, Peptide Array
0.2 mg/ml
Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array
Purified
Mouse
IgG2a
100 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit