Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIM29 Monoclonal Antibody (8C8G5)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA538651
Description
This antibody has been tested in indirect ELISA.
The protein encoded by this gene belongs to the TRIM protein family. It has multiple zinc finger motifs and a leucine zipper motif. It has been proposed to form homo- or heterodimers which are involved in nucleic acid binding. Thus, it may act as a transcriptional regulatory factor involved in carcinogenesis and/or differentiation. It may also function in the suppression of radiosensitivity since it is associated with ataxia telangiectasia phenotype.
Specifications
TRIM29 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q14134 | |
Trim29 | |
Purified recombinant fragment of human TRIM29 (aa: 451-588) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
8C8G5 | |
Unconjugated | |
Trim29 | |
1110047J21Rik; 2810431N19Rik; 4732461M22Rik; AI119726; Ataxia telangiectasia group D-associated protein; ataxia-telangiectasia group D-associated protein; ATDC; EGK_06995; FLJ36085; Trim29; tripartite motif containing 29; tripartite motif protein 29; tripartite motif protein TRIM29; tripartite motif-containing 29; tripartite motif-containing protein 29 | |
Mouse | |
Protein G | |
RUO | |
23650 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction