Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM35 Polyclonal Antibody
TRIM35 Polyclonal Antibody
Supplier: Thermo Scientific PA541561
Description
TRIM35 Polyclonal Antibody for Western Blot

Specifications
TRIM35 | |
Polyclonal | |
Unconjugated | |
TRIM35 | |
0710005M05Rik; A430106H13Rik; AW046487; Hemopoietic lineage switch protein 5; HLS5; KIAA1098; LOW QUALITY PROTEIN: tripartite motif-containing protein 35; macrophage-derived apoptosis-inducing RBCC protein; Mair; MGC17233; mKIAA1098; NC8; Protein MAIR; Protein Nc8; Trim35; tripartite motif containing 35; tripartite motif protein 35; tripartite motif-containing 35; tripartite motif-containing protein 35 | |
Rabbit | |
Protein A | |
RUO | |
23087 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UPQ4 | |
TRIM35 | |
Synthetic peptide directed towards the C-terminal of human TRIM35 (aa 401-450). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction