Learn More
TRIM41 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 184681AP
Catalog No. 50-173-2158
Add to cart
Description
Description
TRIM41 belongs to a family of tripartite motif (TRIM) proteins defined as containing a RING finger, one or more B-box domains, and a coiled-coil region (Tanaka et al., 2005 ). See TRIM45.
Specifications
Specifications
TRIM41
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5NCC3, Q8WV44
RINCK, TRIM41, tripartite motif containing 41
Rabbit
IgG
150 μL
-20°C
Primary
211007, 303088, 90933
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
TRIM41
Liquid
TRIM41
TRIM41 Fusion Protein Ag13236
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat