Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM44 Mouse anti-Human, Clone: OTI1E10, liquid, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene TA505561
Description
May play a role in the process of differentiation and maturation of neuronal cells. May regulate the activity of TRIM17.Specifications
TRIM44 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96DX7 | |
AN3, DIPB, HSA249128, MC7 | |
Mouse | |
IgG1 | |
100 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
54765 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
OTI1E10 | |
Unconjugated | |
TRIM44 | |
Liquid | |
TRIM44 | |
Full length human recombit protein of human TRIM44 produced in HEK293T cell. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title