Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM45 Monoclonal Antibody (OTI3G3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526228
Description
TRIM45 Monoclonal antibody specifically detects TRIM45 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
TRIM45 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H8W5 | |
TRIM45 | |
Human recombinant protein fragment corresponding to amino acids 288-580 of TRIM45 produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
OTI3G3 | |
Unconjugated | |
TRIM45 | |
4921530N01Rik; FLJ13181; RING finger protein 99; RNF99; TRIM45; tripartite motif containing 45; tripartite motif protein 45; tripartite motif-containing 45; tripartite motif-containing protein 45 | |
Mouse | |
Affinity Chromatography | |
RUO | |
80263 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction