Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM47 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550892

Catalog No. PA550892


Add to cart

Description

Description

The antibody detects endogenous levels of total TRIM47 protein.

Gene overexpressed in astrocytoma
Specifications

Specifications

TRIM47
Polyclonal
Unconjugated
TRIM47
Liquid
TRIM47
Synthetic peptide corresponding to a region derived from internal residues of human tripartite motif containing 47
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1.9 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Q96LD4, Q8C0E3
GOA, RNF100
Rabbit
IgG
100 μL
-20°C
Primary
217333, 91107
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit