Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM49 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP155061

 View more versions of this product

Catalog No. NBP155061


Add to cart

Description

Description

TRIM49 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

TRIM49
Unconjugated
PBS & 2% Sucrose. with 0.09% Sodium Azide
Affinity Purified
TRIM49
Synthetic peptides corresponding to TRIM49(tripartite motif-containing 49) The peptide sequence was selected from the N terminal of TRIM49. Peptide sequence RPCFYLNWQDIPFLVQCSECTKSTEQINLKTNIHLKKMASLARKVSLWLF.
53 kDa
100 ul
Zinc Finger
Primary
57093
Human
Western Blot
Western Blot 1:100-1:2000
Q9NS80
RING finger protein 18TRIM49L2, RNF18testis-specific RING Finger protein, Testis-specific RING-finger protein, tripartite motif containing 49, tripartite motif-containing 49, tripartite motif-containing protein 49
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit