Learn More
Aviva Systems Biology Corp TRIM50 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP34494_P050
This is a rabbit polyclonal antibody against TRIM50. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TRIM50. Alternate names: FLJ32804, MGC138357, MGC138359, TRIM50AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55418