Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIM52 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NBP23165125UL
Description
TRIM52 Polyclonal specifically detects TRIM52 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TRIM52 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
Q96A61 | |
TRIM52 | |
This antibody was developed against a recombinant protein corresponding to amino acids: EEVVQEYQEIKLETTLVGILQIEQESIHSKAYNQ | |
25 μL | |
Zinc Finger | |
84851 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
MGC16175, RING finger protein 102, RNF102tripartite motif-containing 52, tripartite motif containing 52, tripartite motif-containing protein 52 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction