Learn More
TRIM54 Mouse anti-Human, Clone: OTI4E7, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF803766
Catalog No. 50-166-6656
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene contains a RING finger motif and is highly similar to the ring finger proteins RNF28/MURF1 and RNF29/MURF2. In vitro studies demonstrated that this protein, RNF28, and RNF29 form heterodimers, which may be important for the regulation of titin kinase and microtubule-dependent signal pathways in striated muscles. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications
Specifications
TRIM54
Monoclonal
Unconjugated
TRIM54
Lyophilized
TRIM54
Human recombit protein fragment corresponding to amino acids 166-400 of human TRIM54 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI4E7
PBS with 8% trehalose and no preservative; pH 7.3
Q9BYV2
MURF, MURF-3, RNF30, muRF3
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
57159