Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM58 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549123

Catalog No. PA549123


Add to cart

Description

Description

E3 ubiquitin ligase induced during late erythropoiesis. Directly binds and ubiquitinates the intermediate chain of the microtubule motor dynein (DYNC1LI1/DYNC1LI2), stimulating the degradation of the dynein holoprotein complex. May participate in the erythroblast enucleation process through regulation of nuclear polarization.
Specifications

Specifications

TRIM58
Polyclonal
Unconjugated
TRIM58
Liquid
TRIM58
KLH conjugated synthetic peptide between 457-486 amino acids from the C-terminal region of human TRIM58.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.3 mg/mL
PBS with 0.09% sodium azide
Q8NG06
E3 ubiquitin-protein ligase TRIM58, 632-, Protein BIA2, Tripartite motif-containing protein 58, TRIM58
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
25893
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit