Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM60 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549378

Catalog No. PA549378

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene contains a RING finger domain, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Pseudogenes of this gene are located on more than six chromosomes including chromosome 4. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq, Jan 2013]
Specifications

Specifications

TRIM60
Polyclonal
PBS with 0.09% sodium azide
Q495X7
Tripartite motif-containing protein 60, RING finger protein 129, RING finger protein 33, TRIM60, RNF129, RNF33
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
166655
Western Blot
Unconjugated
TRIM60
Liquid
TRIM60
KLH conjugated synthetic peptide between 177-211 amino acids from the Central region of human TRIM60.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit