Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TRIM74 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548925

Catalog No. PA548925

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TRIM74
Polyclonal
PBS with 0.09% sodium azide
Q86UV6
Tripartite motif-containing protein 74, Tripartite motif-containing protein 50C, TRIM74, TRIM50C
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
378108
Western Blot
Unconjugated
TRIM74
Liquid
TRIM74
KLH conjugated synthetic peptide between 166-192 amino acids from the C-terminal region of human TRIM74.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit