Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoprim, 50mg/mL in DMSO, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
1mL
5 x 1mL
738-70-5
C14H18N4O3
290.323
MFCD00036761
IEDVJHCEMCRBQM-UHFFFAOYSA-N
5578
CHEBI:45924
5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N
738-70-5
C14H18N4O3
290.323
MFCD00036761
IEDVJHCEMCRBQM-UHFFFAOYSA-N
5578
CHEBI:45924
5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N
Packaging Plastic bottle
Physical Form Liquid
Quantity 1mL
Chemical Name or Material Trimethoprim