Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoprim, 50mg/mL in DMSO, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
1mL
5 x 1mL
738-70-5
C14H18N4O3
290.323
MFCD00036761
IEDVJHCEMCRBQM-UHFFFAOYSA-N
5578
CHEBI:45924
5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N
738-70-5
C14H18N4O3
290.323
MFCD00036761
IEDVJHCEMCRBQM-UHFFFAOYSA-N
5578
CHEBI:45924
5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N
Physical Form Liquid
Quantity 5 x 1mL
Chemical Name or Material Trimethoprim, 50mg/mL in DMSO, sterile-filtered

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UV8225000

storageNote1 : -30° to -10°C