Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoprim lactate, 20mg/mL in distilled water, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
1mL
5 x 1mL
23256-42-0
C17H24N4O6
380.401
MFCD00171722
IIZVTUWSIKTFKO-UHFFFAOYSA-N
3084396
2-hydroxypropanoic acid;5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
CC(C(=O)O)O.COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N
23256-42-0
C17H24N4O6
380.401
MFCD00171722
IIZVTUWSIKTFKO-UHFFFAOYSA-N
3084396
2-hydroxypropanoic acid;5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
CC(C(=O)O)O.COC1=CC(=CC(=C1OC)OC)CC2=CN=C(N=C2N)N
Physical Form Liquid
Quantity 5 x 1mL
Chemical Name or Material Trimethoprim lactate, 20mg/mL in distilled water, sterile-filtered

storageNote1 : -30° to -10°C