Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoprim, Micronized, USP, 98.5-101%, Spectrum™

Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum

T1228, 738-70-5, C14H18N4O3

Spectrum™ Trimethoprim, Micronized, USP is an antibiotic used to treat bacterial infections. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS 738-70-5
CAS Max % 1
Ignition Residue 0.001
Molecular Formula C14H18N4O3
Loss on Drying 0.005
Packaging Fiber Drum
InChI Key IEDVJHCEMCRBQM-UHFFFAOYSA-N
SMILES COC1=CC(CC2=CN=C(N)N=C2N)=CC(OC)=C1OC
IUPAC Name 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
Molecular Weight (g/mol) 290.32
Percent Purity 98.5 to 101%
Grade USP
Quantity 25 kg
Show More Show Less