Alfa Aesar™ Trimethoxyboroxine

Manufacturer: Alfa Aesar™ 8915318

Quantity 50g

 View more versions of this product

Catalog No. AA8915318

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 102-24-9
    Molecular Formula (CH3O)3B3O3
    Formula Weight 173.53
    MDL Number MFCD00005946
    UN Number UN2924
    Beilstein 1709158
    Chemical Name or Material Trimethoxyboroxine

    Specifications

    Formula Weight 173.53
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    Health Hazard 2 GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Quantity 50g
    CAS 102-24-9
    UN Number UN2924
    Beilstein 1709158
    Health Hazard 1 Danger
    Flash Point 14°C (57°F)
    Refractive Index 1.3996
    Molecular Formula (CH3O)3B3O3
    MDL Number MFCD00005946
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    RTECS Number ED8720000
    Density 1.216g/mL
    Odor Strong
    Sensitivity Air & Moisture Sensitive
    Boiling Point 130°C
    Melting Point 10° to 11°C
    TSCA TSCA
    Chemical Name or Material Trimethoxyboroxine

    GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit