Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethoxyboroxine, ACROS Organics™

Click to view available options
Quantity:
25g
100g
250g
Packaging:
Glass bottle
102-24-9
C3H9B3O6
173.529
MFCD00005946
ZFMOJHVRFMOIGF-UHFFFAOYSA-N
trimethoxyboroxine,trimethoxyboroxin,boroxin, trimethoxy,trimethoxycyclotriboroxane,2,4,6-trimethoxyboroxin,triboron trimethyl hexaoxide,boroxin, 2,4,6-trimethoxy,boric acid h3b3o6 , trimethyl ester,trimethoxy-boroxi,trimethoxyboroxole
66880
2,4,6-trimethoxy-1,3,5,2,4,6-trioxatriborinane
B1(OB(OB(O1)OC)OC)OC
102-24-9
C3H9B3O6
173.529
MFCD00005946
ZFMOJHVRFMOIGF-UHFFFAOYSA-N
trimethoxyboroxine,trimethoxyboroxin,boroxin, trimethoxy,trimethoxycyclotriboroxane,2,4,6-trimethoxyboroxin,triboron trimethyl hexaoxide,boroxin, 2,4,6-trimethoxy,boric acid h3b3o6 , trimethyl ester,trimethoxy-boroxi,trimethoxyboroxole
66880
2,4,6-trimethoxy-1,3,5,2,4,6-trioxatriborinane
B1(OB(OB(O1)OC)OC)OC
Color Colorless
Packaging Glass bottle
Quantity 100g
Density 1.2160g/mL
Formula Weight 173.53
Physical Form Liquid
Percent Purity 18 to 19.5% (as B)
Chemical Name or Material Trimethoxyboroxine

GHS H Statement:
Causes serious eye damage.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger