Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl 1,2,4-benzenetricarboxylate, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
2459-10-1
98%
C12H12O6
C6H3(CO2CH3)3
MFCD00043629
Trimethyl 1, 2, 4-benzenetricarboxylate, 98%
2459-10-1
98%
C12H12O6
C6H3(CO2CH3)3
MFCD00043629
Trimethyl 1, 2, 4-benzenetricarboxylate, 98%
Infrared Spectrum Authentic
Density 1.26g/mL
Quantity 10g
Refractive Index 1.5220 to 1.5245 (20°C, 589nm)
Color Colorless to Yellow
Beilstein 09, I, 429
Synonym trimethyl trimellitate,trimethyl 1,2,4-benzenetricarboxylate,methyl trimellitate,trimellitic acid trimethyl ester,1,2,4-benzenetricarboxylic acid, trimethyl ester,unii-16o7dt5n66,1,2,4-benzenetricarboxylic acid trimethyl ester,1,2,4-benzenetricarboxylic acid, 1,2,4-trimethyl ester,acmc-1ckkm,dsstox_cid_24620
InChI Key MJHNUUNSCNRGJE-UHFFFAOYSA-N
Molecular Weight (g/mol) 252.22
PubChem CID 17159
Formula Weight 252.22
Physical Form Viscous Liquid
Percent Purity ≥97.5%
Show More Show Less