Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Trimethyl borate, 70% azeotrope with methanol

Manufacturer:  Alfa Aesar™ 44721K2

Quantity 1L

 View more versions of this product

Catalog No. AA44721K2


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

121-43-7
103.92
UN1993
Trimethyl borate
(CH3O)3B
MFCD00008346
14,9712

Specifications

70% azeotrope with methanol
−9°C (16°F)
52° to 54°C
121-43-7
(CH3O)3B
MFCD00008346
14,9712
0.885g/mL
Characteristic
1L
Liquid
103.92
UN1993
Trimethyl borate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H300-H330-H370-H226-H312
Fatal if swallowed.
Fatal if inhaled.
Causes damage to organs.
Flammable liquid and vapor.
Harmful in contact with skin.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit