Alfa Aesar™ Trimethyl borate, 70% azeotrope with methanol

Manufacturer: Alfa Aesar™ 44721AK

Quantity 250mL

 View more versions of this product

Catalog No. AA44721AK

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 121-43-7
    Molecular Formula (CH3O)3B
    Formula Weight 103.92
    MDL Number MFCD00008346
    UN Number UN1993
    Merck Index 14,9712
    Chemical Name or Material Trimethyl borate

    Specifications

    Formula Weight 103.92
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    CAS 121-43-7
    Density 0.885g/mL
    Odor Characteristic
    Boiling Point 52° to 54°C
    Quantity 250mL
    UN Number UN1993
    Merck Index 14,9712
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Concentration 70% azeotrope with methanol
    Physical Form Liquid
    Molecular Formula (CH3O)3B
    MDL Number MFCD00008346
    Health Hazard 2 GHS H Statement
    H300-H330-H370-H226-H312
    Fatal if swallowed.
    Fatal if inhaled.
    Causes damage to organs.
    Flammable liquid and vapor.
    Harmful in contact with skin.
    Flash Point −9°C (16°F)
    Chemical Name or Material Trimethyl borate

    GHS H Statement
    H300-H330-H370-H226-H312
    Fatal if swallowed.
    Fatal if inhaled.
    Causes damage to organs.
    Flammable liquid and vapor.
    Harmful in contact with skin.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit