Alfa Aesar™ Trimethyl borate, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 4376536

Quantity 500g

 View more versions of this product

Catalog No. AA4376536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 121-43-7
    Molecular Formula (CH3O)3B
    Formula Weight 103.92
    MDL Number MFCD00008346
    UN Number UN2416
    Beilstein 1697939
    Merck Index 14,9712
    Chemical Name or Material Trimethyl borate

    Specifications

    Formula Weight 103.92
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Physical Form Liquid
    Molecular Formula (CH3O)3B
    MDL Number MFCD00008346
    CAS 121-43-7
    Beilstein 1697939
    Density 0.915g/mL
    Flash Point −8°C (17°F)
    Boiling Point 67° to 69°C
    Quantity 500g
    TSCA TSCA
    Merck Index 14,9712
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIMETHYL BORATE
    Refractive Index 1.3610
    Health Hazard 2 GHS H Statement
    H225-H312
    Highly flammable liquid and vapor.
    Harmful in contact with skin.
    RTECS Number ED5600000
    Sensitivity Moisture Sensitive
    Melting Point −34°C
    UN Number UN2416
    Odor Odorless
    Color Colorless
    Chemical Name or Material Trimethyl borate
    Assay 99.99%, (metals basis)

    GHS H Statement
    H225-H312
    Highly flammable liquid and vapor.
    Harmful in contact with skin.

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit