Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl borate, 99.9995+% (metals basis)

Manufacturer:  Alfa Aesar 2297509

Quantity 10g

 View more versions of this product

Catalog No. AA2297509


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

121-43-7
(CH3O)3B
MFCD00008346
1697939
Trimethyl borate
C3H9BO3
103.92
UN2416
14,9712

Specifications

121-43-7
>99.9995% (metals basis)
(CH3O)3B
MFCD00008346
1697939
0.915g/mL
Trimethyl borate
Liquid
C3H9BO3
103.92
UN2416
14,9712
10g
≥99.9995%, (metals basis)

Safety and Handling

SDS & Safety Documents

GHS H Statement
H225-H312

GHS P Statement
P210-P280-P240-P241-P233-P303+P361+P353-P312-P363-P403+P235-P501a

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TRIMETHYL BORATE

RTECSNumber : ED5600000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit