Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl borate, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
121-43-7
C3H9O3B
103.91
MFCD00008346
WRECIMRULFAWHA-UHFFFAOYSA-N
trimethoxyborane,methyl borate,trimethoxyboron,boric acid, trimethyl ester,trimethylborate,boric acid trimethyl ester,trimethoxyborine,borester o,boric acid h3bo3 , trimethyl ester,b och3 3
8470
CHEBI:38913
trimethyl borate
B(OC)(OC)OC
121-43-7
C3H9O3B
103.91
MFCD00008346
WRECIMRULFAWHA-UHFFFAOYSA-N
trimethoxyborane,methyl borate,trimethoxyboron,boric acid, trimethyl ester,trimethylborate,boric acid trimethyl ester,trimethoxyborine,borester o,boric acid h3bo3 , trimethyl ester,b och3 3
8470
CHEBI:38913
trimethyl borate
B(OC)(OC)OC
Viscosity 0.36 mPa.s (25°C)
Specific Gravity 0.915
Infrared Spectrum Authentic
Refractive Index 1.3562 to 1.3582
Density 0.9150g/mL
Quantity 2.5kg
Flash Point −8°C
Boiling Point 68.0°C to 69.0°C
Packaging Glass bottle
Color Colorless
Melting Point -34.0°C
Assay Percent Range 99%
Linear Formula (CH3O)3B
Beilstein 01, 287
Fieser 01,673; 13,218
Merck Index 15, 9882
Solubility Information Solubility in water: reacts. Other solubilities: miscible with thf,diethyl ether,hexane,methanol,,isopropylamine,and other organic liquids
Formula Weight 103.91
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Trimethyl borate, 99%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 204-468-9

RTECSNumber : ED5600000

TSCA : TSCA