Learn More
trimethyl-Histone H3 (Lys4) Rabbit anti-Human,Mouse, BSA, Clone: MC315, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB047914X25UL
 View more versions of this product
Catalog No. ZRB047914MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
trimethyl-Histone H3 (Lys4) Monoclonal antibody specifically detects trimethyl-Histone H3 (Lys4) in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Peptide Inhibition Assay, Western Blotting.
Specifications
Specifications
trimethyl-Histone H3 (Lys4)
MC315
BSA
Histone H3Kme3;Histone H3;Trimethylated Histone H3
Rabbit
IgG
Protein A Purified
Epigenetics and Nuclear Function
Primary
4X
Immunocytochemistry, Immunohistochemistry, Inhibition Assays, Western Blot
30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Purified
HIST3H3, H3FT
BSA-conjugated linear peptide corresponding to an amino acid sequence containing (RTK4me3Q) (trimethylated lysine 4) from human Histone H3.
∼17 kDa observed; 15.51 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
4 x 25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Human, Mouse