Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trimethyl-Histone H3 (Lys4) Rabbit anti-Human,Mouse, BSA, Clone: MC315, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen trimethyl-Histone H3 (Lys4)
Applications Immunocytochemistry, Immunohistochemistry, Inhibition Assays, Western Blot
Conjugate BSA
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB047912MI
View Documents
MilliporeSigma
ZRB0479125UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB047914MI
View Documents
MilliporeSigma
ZRB047914X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

trimethyl-Histone H3 (Lys4) Monoclonal antibody specifically detects trimethyl-Histone H3 (Lys4) in Human,Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry, Peptide Inhibition Assay, Western Blotting.
Specifications

Specifications

trimethyl-Histone H3 (Lys4)
BSA
Rabbit
Histone H3Kme3;Histone H3;Trimethylated Histone H3
BSA-conjugated linear peptide corresponding to an amino acid sequence containing (RTK4me3Q) (trimethylated lysine 4) from human Histone H3.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry, Inhibition Assays, Western Blot
Purified
Human, Mouse
HIST3H3, H3FT
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit