Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Trimethyl)methylcyclopentadienylplatinum(IV), 99%, ACROS Organics™

Click to view available options
Quantity:
100mg
Packaging:
Glass bottle
94442-22-5
C9H16Pt
MFCD00079665
94442-22-5
C9H16Pt
MFCD00079665
Quantity 100mg
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Melting Point 29°C to 32°C
NMR 98.5% min.
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 319.32
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (Trimethyl)methylcyclopentadienylplatinum(IV)
Show More Show Less

AIR SENSITIVE

TSCA : TSCA