Alfa Aesar™ Trimethyl orthobenzoate, 98%

Manufacturer: Alfa Aesar™ A1027018

Quantity 50g

 View more versions of this product

Catalog No. AAA1027018

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 707-07-3
    Molecular Formula (C6H5)C(OCH3)3
    Formula Weight 182.22
    MDL Number MFCD00008474
    Beilstein 777623
    Synonym Orthobenzoic acid trimethyl ester; alpha,alpha,alpha-Trimethoxytoluene
    Chemical Name or Material Trimethyl orthobenzoate

    Specifications

    Formula Weight 182.22
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula (C6H5)C(OCH3)3
    MDL Number MFCD00008474
    Beilstein 777623
    Quantity 50g
    CAS 707-07-3
    Synonym Orthobenzoic acid trimethyl ester; alpha,alpha,alpha-Trimethoxytoluene
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Boiling Point 87° to 88°C (7mmHg)
    TSCA TSCA
    Flash Point 82°C (179°F)
    Chemical Name or Material Trimethyl orthobenzoate
    Assay 98%

    Description

    Applications
    Trimethyl orthobenzoate was used in the preparation of 5-phenyl-4,6-dipyrrin and 2,3-orthoester derivatives of ethyl-1-thio-α-L-rhamnopyranoside. It reacts with indole to prepare indol-3-yl-phenyl-methanone.

    Solubility
    Soluble in organic solvents.

    Notes
    Moisture Sensitive. Protect from humidity and water. Store under dry inert gas. Incompatible with oxidizing agents and moisture.

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit