Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl orthobutyrate 97%, ACROS Organics™

Click to view available options
Quantity:
10mL
50mL
Packaging:
Glass bottle
43083-12-1
C7H16O3
148.202
MFCD00008480
JAFMOTJMRSZOJE-UHFFFAOYSA-N
trimethyl orthobutyrate,trimethylorthobutyrate,butane, 1,1,1-trimethoxy,ortho-n-butyric acid trimethyl ester,orthobutyric acid trimethyl ester,orthobutyricacidtrimethylester,1,1,1-trimethoxybutan,trimethy orthobutyrate,pubchem21070,acmc-1aqt6
123512
1,1,1-trimethoxybutane
CCCC(OC)(OC)OC
43083-12-1
C7H16O3
148.202
MFCD00008480
JAFMOTJMRSZOJE-UHFFFAOYSA-N
trimethyl orthobutyrate,trimethylorthobutyrate,butane, 1,1,1-trimethoxy,ortho-n-butyric acid trimethyl ester,orthobutyric acid trimethyl ester,orthobutyricacidtrimethylester,1,1,1-trimethoxybutan,trimethy orthobutyrate,pubchem21070,acmc-1aqt6
123512
1,1,1-trimethoxybutane
CCCC(OC)(OC)OC
Quantity 10mL
Color Colorless to Yellow
Refractive Index 1.4020 to 1.4040 (20°C, 589nm)
Infrared Spectrum Authentic
Density 0.932g/mL
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula CH3(CH2)2C(OCH3)3
Formula Weight 148.2
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Trimethyl orthobutyrate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning