Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Trimethyl)pentamethylcyclopentadienyltitanium(IV) ≥97%, Acros Organics

Click to view available options
Quantity:
1g
107333-47-1
C13H29Ti-5
233.242
MFCD00269851
DVHMCUKDWMNRGX-UHFFFAOYSA-N
acmc-1bo0g,titanium,trimethyl 1,2,3,4,5-h-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl
57346580
carbanide;cyclopenta-1,3-diene;titanium(4+)
[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].C1C=CC=[C-]1.[Ti+4]
107333-47-1
C13H29Ti-5
233.242
MFCD00269851
DVHMCUKDWMNRGX-UHFFFAOYSA-N
acmc-1bo0g,titanium,trimethyl 1,2,3,4,5-h-1,2,3,4,5-pentamethyl-2,4-cyclopentadien-1-yl
57346580
carbanide;cyclopenta-1,3-diene;titanium(4+)
[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].[CH3-].C1C=CC=[C-]1.[Ti+4]
Quantity 1g
Color Yellow
Infrared Spectrum Authentic
Elemental Analysis 97.0% min.
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97+%
Formula Weight 228.21
Physical Form Crystals
Chemical Name or Material (Trimethyl)pentamethylcyclopentadienyltitanium(IV)