Alfa Aesar™ Trimethyl phosphite, 99%

Manufacturer: Alfa Aesar™ 4398818

Quantity 50g

 View more versions of this product

Catalog No. AA4398818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 121-45-9
    Molecular Formula C3H9O3P
    Linear Formula (CH3O)3P
    Formula Weight 124.08
    MDL Number MFCD00008350
    UN Number UN2329
    Chemical Name or Material Trimethyl phosphite

    Specifications

    Formula Weight 124.08
    Health Hazard 2 GHS H Statement
    H226-H361-H302-H315-H319-H335
    Flammable liquid and vapor.
    Suspected of damaging fertility or the unborn child.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Unpleasant
    Molecular Formula C3H9O3P
    Density 1.052g/mL
    UN Number UN2329
    Health Hazard 1 Warning
    Flash Point 27°C (80°F)
    CAS 121-45-9
    Sensitivity Air & Moisture Sensitive
    Boiling Point 111° to 112°C
    TSCA TSCA
    Refractive Index 1.4080
    Linear Formula (CH3O)3P
    MDL Number MFCD00008350
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TRIMETHYL PHOSPHITE
    Color Colorless
    Melting Point −78°C
    Quantity 50g
    Physical Form Liquid
    Chemical Name or Material Trimethyl phosphite
    Assay 99%

    GHS H Statement
    H226-H361-H302-H315-H319-H335
    Flammable liquid and vapor.
    Suspected of damaging fertility or the unborn child.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit