Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl phosphonoacetate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
5927-18-4
C5H11O5P
182.11
SIGOIUCRXKUEIG-UHFFFAOYSA-N
trimethyl phosphonoacetate,methyl 2-dimethoxyphosphoryl acetate,trimethyl phosphonacetate,trimethylacetyl phosphate,acetic acid, dimethoxyphosphinyl-, methyl ester,methyl-dimethoxyphosphoryl acetat,trimethyl phosphoroacetate,phosphonoacetic acid trimethyl ester,methyl dimethylphosphonoacetate,methyl dimethoxyphosphoryl acetate
80029
methyl 2-dimethoxyphosphorylacetate
COC(=O)CP(=O)(OC)OC
5927-18-4
C5H11O5P
182.11
SIGOIUCRXKUEIG-UHFFFAOYSA-N
trimethyl phosphonoacetate,methyl 2-dimethoxyphosphoryl acetate,trimethyl phosphonacetate,trimethylacetyl phosphate,acetic acid, dimethoxyphosphinyl-, methyl ester,methyl-dimethoxyphosphoryl acetat,trimethyl phosphoroacetate,phosphonoacetic acid trimethyl ester,methyl dimethylphosphonoacetate,methyl dimethoxyphosphoryl acetate
80029
methyl 2-dimethoxyphosphorylacetate
COC(=O)CP(=O)(OC)OC
For C-C Bond Formation (Olefination)
Density 1.2600g/mL
Quantity 100g
Color Colorless to Yellow
Refractive Index 1.4360 to 1.4380
Flash Point >112°C
Boiling Point 265.0°C to 268.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Specific Gravity 1.26
Assay Percent Range 98%
Linear Formula (CH3O)2P(O)CH2CO2CH3
Fieser 02,442; 07,394; 01,314; 02,169; 01,1236; 02,171; 13,15
Formula Weight 182.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Trimethyl phosphonoacetate
Show More Show Less