Alfa Aesar™ Trimethyl thiophosphate

Manufacturer: Alfa Aesar™ 3039514

Quantity 25g

 View more versions of this product

Catalog No. AA3039514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 152-18-1
    Formula Weight 156.15
    MDL Number MFCD00014888
    UN Number UN3278
    Chemical Name or Material Trimethyl thiophosphate

    Specifications

    Formula Weight 156.15
    Health Hazard 3 GHS P Statement
    P260-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H330-H302-H315-H319-H335
    Fatal if inhaled.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    UN Number UN3278
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
    Color Off-white
    Quantity 25g
    Odor Sulphurous
    Physical Form Liquid
    MDL Number MFCD00014888
    CAS 152-18-1
    Health Hazard 1 Danger
    Chemical Name or Material Trimethyl thiophosphate

    GHS H Statement
    H330-H302-H315-H319-H335
    Fatal if inhaled.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P260-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit