Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylacetamide 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Plastic bottle
754-10-9
C5H11NO
101.15
MFCD00008011
XIPFMBOWZXULIA-UHFFFAOYSA-N
pivalamide,trimethylacetamide,2,2,2-trimethylacetamide,propanamide, 2,2-dimethyl,2,2-dimethylpropionamide,2,2-dimethyl-propionamide,unii-fes86mr7zi,fes86mr7zi,propanamide,2,2-dimethyl,trimethylacetarnide
12957
2,2-dimethylpropanamide
CC(C)(C)C(=O)N
754-10-9
C5H11NO
101.15
MFCD00008011
XIPFMBOWZXULIA-UHFFFAOYSA-N
pivalamide,trimethylacetamide,2,2,2-trimethylacetamide,propanamide, 2,2-dimethyl,2,2-dimethylpropionamide,2,2-dimethyl-propionamide,unii-fes86mr7zi,fes86mr7zi,propanamide,2,2-dimethyl,trimethylacetarnide
12957
2,2-dimethylpropanamide
CC(C)(C)C(=O)N
Boiling Point 212°C
Quantity 5g
Infrared Spectrum Authentic
Color White to Yellow
Melting Point 152°C to 156°C
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula (CH3)3CCONH2
Beilstein 02, 320
Formula Weight 101.15
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Trimethylacetamide, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 212-043-4

TSCA : TSCA