Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylacetic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic drum
75-98-9
C5H10O2
102.13
MFCD00004194
IUGYQRQAERSCNH-UHFFFAOYSA-N
pivalic acid,trimethylacetic acid,neopentanoic acid,2,2-dimethylpropionic acid,tert-pentanoic acid,propanoic acid, 2,2-dimethyl,versatic 5,kyselina pivalova,acetic acid, trimethyl,alpha,alpha-dimethylpropionic acid
6417
CHEBI:45133
2,2-dimethylpropanoic acid
CC(C)(C)C(=O)O
75-98-9
C5H10O2
102.13
MFCD00004194
IUGYQRQAERSCNH-UHFFFAOYSA-N
pivalic acid,trimethylacetic acid,neopentanoic acid,2,2-dimethylpropionic acid,tert-pentanoic acid,propanoic acid, 2,2-dimethyl,versatic 5,kyselina pivalova,acetic acid, trimethyl,alpha,alpha-dimethylpropionic acid
6417
CHEBI:45133
2,2-dimethylpropanoic acid
CC(C)(C)C(=O)O
Viscosity 3.6 mm2/s (60°C)
Melting Point 33.0°C to 35.0°C
Boiling Point 163.0°C to 164.0°C
Color Tan to Yellow
Quantity 100g
Infrared Spectrum Authentic
Packaging Glass bottle
Flash Point 63°C
Assay Percent Range 99%
Linear Formula (CH3)3CCO2H
Beilstein 02, 319
Fieser 01,1229
Merck Index 15, 7625
Solubility Information Solubility in water: 25g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 102.13
Physical Form Powder, Crystals and/or Chunks
Percent Purity 99%
Chemical Name or Material Trimethylacetic acid, 99%
Show More Show Less

GHS P Statement
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-922-5

RTECSNumber : TO7700000

TSCA : TSCA