Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylacetonitrile, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
630-18-2
C5H9N
83.13
MFCD00001847
JAMNHZBIQDNHMM-UHFFFAOYSA-N
pivalonitrile,trimethylacetonitrile,2,2-dimethylpropionitrile,tert-butyl cyanide,propanenitrile, 2,2-dimethyl,tert-butylnitrile,2-cyano-2-methylpropane,trimethyl acetonitrile,tert-cyanobutane,t-bucn
12416
2,2-dimethylpropanenitrile
CC(C)(C)C#N
630-18-2
C5H9N
83.13
MFCD00001847
JAMNHZBIQDNHMM-UHFFFAOYSA-N
pivalonitrile,trimethylacetonitrile,2,2-dimethylpropionitrile,tert-butyl cyanide,propanenitrile, 2,2-dimethyl,tert-butylnitrile,2-cyano-2-methylpropane,trimethyl acetonitrile,tert-cyanobutane,t-bucn
12416
2,2-dimethylpropanenitrile
CC(C)(C)C#N
Melting Point 15°C to 16°C
Boiling Point 105°C to 106°C
Flash Point 4°C
Refractive Index 1.3765 to 1.3785
Specific Gravity 0.75
Quantity 25mL
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Density 0.7500g/mL
Assay Percent Range 98%
Linear Formula (CH3)3CCN
Beilstein 02, 320
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,acetone and toluene
Formula Weight 83.13
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Trimethylacetonitrile
Show More Show Less

GHS P Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger