Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylaluminium, 1.0M solution in heptane, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
75-24-1,142-82-5
C3H9Al
72.087
MFCD00008252
JLTRXTDYQLMHGR-UHFFFAOYSA-N
trimethylaluminum,trimethylaluminium,aluminum, trimethyl,trimethylalane,unii-av210lg46j,ch3 3al,aluminum trimethanide,aluminum trimethyl,trimethylaluminum solution,trimethylaluminum, elec. gr.
16682925
trimethylalumane
C[Al](C)C
75-24-1,142-82-5
C3H9Al
72.087
MFCD00008252
JLTRXTDYQLMHGR-UHFFFAOYSA-N
trimethylaluminum,trimethylaluminium,aluminum, trimethyl,trimethylalane,unii-av210lg46j,ch3 3al,aluminum trimethanide,aluminum trimethyl,trimethylaluminum solution,trimethylaluminum, elec. gr.
16682925
trimethylalumane
C[Al](C)C
Lewis acid
Linear Formula (CH3)3Al
Fieser 08,506; 14,46; 17,372
Merck Index 15, 9879
Density 0.6880g/mL
Solubility Information Solubility in water: reacts. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons
Formula Weight 72.09
Physical Form Liquid
Percent Purity 9.5 to 10.5 wt% (on Al basis)
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: reacts. Other solubilities: miscible with saturated aliphatic and aromatic, hydrocarbons
Specific Gravity 0.688
Viscosity 0.90 mPa.s (30°C)
Boiling Point 126.0°C
Color Colorless
Melting Point 15.0°C
Quantity 100mL
Flash Point −4°C
Chemical Name or Material Trimethylaluminium, 1.0M Solution in Heptane
Show More Show Less

GHS H Statement:
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger