Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylaluminum (15% in Toluene, ca. 1.8mol/L), TCI America™

Click to view available options
Quantity:
100mL
75-24-1
C3H9Al
72.087
MFCD00008252
JLTRXTDYQLMHGR-UHFFFAOYSA-N
trimethylaluminum,trimethylaluminium,aluminum, trimethyl,trimethylalane,unii-av210lg46j,ch3 3al,aluminum trimethanide,aluminum trimethyl,trimethylaluminum solution,trimethylaluminum, elec. gr.
16682925
trimethylalumane
C[Al](C)C
75-24-1
C3H9Al
72.087
MFCD00008252
JLTRXTDYQLMHGR-UHFFFAOYSA-N
trimethylaluminum,trimethylaluminium,aluminum, trimethyl,trimethylalane,unii-av210lg46j,ch3 3al,aluminum trimethanide,aluminum trimethyl,trimethylaluminum solution,trimethylaluminum, elec. gr.
16682925
trimethylalumane
C[Al](C)C
UN Number 3394
Formula Weight 72.09
Physical Form Clear Liquid at 20°C
Color Colorless
Quantity 100mL
Chemical Name or Material Trimethylaluminum (15% in Toluene, ca. 1.8mol/L)

EINECSNumber : (2)-2227

RTECSNumber : BD2204000

TSCA : Yes