Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylamine, 1M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
75-50-3,109-99-9
C3H9N
59.112
MFCD00008327
GETQZCLCWQTVFV-UHFFFAOYSA-N
trimethylamine,methanamine, n,n-dimethyl,dimethylmethaneamine,n-trimethylamine,trimethylamine solution,ch3 3n,trimethyl amine,trimethylamin,trimethyl-amine,fema number 3241
1146
CHEBI:18139
N,N-dimethylmethanamine
CN(C)C
75-50-3,109-99-9
C3H9N
59.112
MFCD00008327
GETQZCLCWQTVFV-UHFFFAOYSA-N
trimethylamine,methanamine, n,n-dimethyl,dimethylmethaneamine,n-trimethylamine,trimethylamine solution,ch3 3n,trimethyl amine,trimethylamin,trimethyl-amine,fema number 3241
1146
CHEBI:18139
N,N-dimethylmethanamine
CN(C)C
Linear Formula N(CH3)3
Concentration 1M, exact strength on the certificate of analysis
Density 0.8500g/mL
Formula Weight 59.11
Physical Form Liquid
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: soluble. Other solubilities: soluble in most organic solvents
Specific Gravity 0.85
Color Colorless to Yellow
Quantity 800mL
Flash Point −17°C
Chemical Name or Material Trimethylamine, 1M solution in THF
Show More Show Less

GHS H Statement
Highly flammable liquid and vapor. Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. Suspected of causing cancer. Harmful if swallowed. May cause drowsiness or dizziness. May form explosive peroxides.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

Danger