Learn More
Trimethylamine, pure, 4.2M (33 wt.%) solution in ethanol, Thermo Scientific Chemicals
$624.00
Specifications
Chemical Name or Material | Trimethylamine |
---|---|
CAS | 75-50-3, 64-17-5 |
Color | Colorless |
Density | 0.7500g/mL |
Boiling Point | 50.0°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AC270440250
|
Thermo Scientific Chemicals
270440250 |
25 mL |
Each for $45.25
Order in Bulk
Request bulk or custom formats |
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
AC270440050
|
Thermo Scientific Chemicals
270440050 |
5 L |
Each for $624.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trimethylamine | |
Colorless | |
50.0°C | |
C3H9N | |
1.3630 to 1.3660 | |
04,43 | |
0.75 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol,ether,benzene,toluene,,xylene,ethylbenzene and chloroform | |
CN(C)C | |
1146 | |
59.11 | |
Pure |
75-50-3, 64-17-5 | |
0.7500g/mL | |
−20°C | |
(CH3)3N | |
MFCD00008327 | |
15,988 | |
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241 | |
GETQZCLCWQTVFV-UHFFFAOYSA-N | |
59.11 | |
CHEBI:18139 | |
32 to 34% | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Highly flammable liquid and vapor.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 200-875-0
RTECSNumber : PA0350000
TSCA : TSCA
For Manufacturing and Laboratory Use. Not for beverage use
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.