Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution, ACROS Organics™

Click to view available options
Quantity:
500mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
75-50-3,7732-18-5
C3H9N
59.112
GETQZCLCWQTVFV-UHFFFAOYSA-N
trimethylamine,methanamine, n,n-dimethyl,dimethylmethaneamine,n-trimethylamine,trimethylamine solution,ch3 3n,trimethyl amine,trimethylamin,trimethyl-amine,fema number 3241
1146
CHEBI:18139
N,N-dimethylmethanamine
CN(C)C
75-50-3,7732-18-5
C3H9N
59.112
GETQZCLCWQTVFV-UHFFFAOYSA-N
trimethylamine,methanamine, n,n-dimethyl,dimethylmethaneamine,n-trimethylamine,trimethylamine solution,ch3 3n,trimethyl amine,trimethylamin,trimethyl-amine,fema number 3241
1146
CHEBI:18139
N,N-dimethylmethanamine
CN(C)C
Linear Formula (CH3)3N
Density 0.8600g/mL
Formula Weight 59.11
Physical Form Liquid
Percent Purity 48 to 52% (total base)
Grade Pure
Packaging Glass bottle
Solubility Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether, benzene, toluene,, xylene, ethylbenzene and chloroform
Specific Gravity 0.86
Boiling Point 30.0°C to 100.0°C
Color Colorless
Melting Point -2.0°C
Quantity 500mL
Flash Point −45°C
Chemical Name or Material Trimethylamine
Show More Show Less