Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyle1,3,5-benzenetricarboxylate, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
250g
Packaging:
Glass bottle
2672-58-4
C12H12O6
252.22
MFCD00008434
RGCHNYAILFZUPL-UHFFFAOYSA-N
trimethyl 1,3,5-benzenetricarboxylate,trimethyl trimesate,trimesic acid trimethyl ester,1,3,5-benzenetricarboxylic acid, trimethyl ester,1,3,5-benzenetricarboxylic acid trimethyl ester,trimethylbenzene-1,3,5-tricarboxylate,1,3,5-trimethyl benzene-1,3,5-tricarboxylate,methyl 3,5-bis methoxycarbonyl benzoate,benzene-1,3,5-tricarboxylic acid trimethyl ester,timtec-bb sbb007766
75881
trimethyl benzene-1,3,5-tricarboxylate
COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC
2672-58-4
C12H12O6
252.22
MFCD00008434
RGCHNYAILFZUPL-UHFFFAOYSA-N
trimethyl 1,3,5-benzenetricarboxylate,trimethyl trimesate,trimesic acid trimethyl ester,1,3,5-benzenetricarboxylic acid, trimethyl ester,1,3,5-benzenetricarboxylic acid trimethyl ester,trimethylbenzene-1,3,5-tricarboxylate,1,3,5-trimethyl benzene-1,3,5-tricarboxylate,methyl 3,5-bis methoxycarbonyl benzoate,benzene-1,3,5-tricarboxylic acid trimethyl ester,timtec-bb sbb007766
75881
trimethyl benzene-1,3,5-tricarboxylate
COC(=O)C1=CC(=CC(=C1)C(=O)OC)C(=O)OC
Melting Point 144°C to 147°C
Quantity 100g
Color White
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula C6H3(CO2CH3)3
Beilstein 09, 979
Formula Weight 252.22
Physical Form Crystalline Powder or Crystals
Percent Purity ≥98.5%
Chemical Name or Material Trimethyl 1,3,5-benzenetricarboxylate
Show More Show Less