Alfa Aesar™ Trimethylene borate, 98%

Manufacturer: Alfa Aesar™ B2193306

Quantity 5g

 View more versions of this product

Catalog No. AAB2193306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 20905-35-5
    Molecular Formula C9H18B2O6
    Formula Weight 243.86
    MDL Number MFCD03701690
    Beilstein 1913020
    Synonym Boric acid cyclic ester with 1,3-propanediol; Boric acid 1,3-propanediol cyclic ester
    Chemical Name or Material Trimethylene borate

    Specifications

    Formula Weight 243.86
    Molecular Formula C9H18B2O6
    MDL Number MFCD03701690
    CAS 20905-35-5
    Beilstein 1913020
    Synonym Boric acid cyclic ester with 1,3-propanediol; Boric acid 1,3-propanediol cyclic ester
    Flash Point >110°C (230°F)
    Boiling Point 145° to 148°C (0.2mmHg)
    Quantity 5g
    Chemical Name or Material Trimethylene borate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit