Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylene oxide, 97%, ACROS Organics™

Click to view available options
Quantity:
25mL
Packaging:
Glass bottle
503-30-0
C3H6O
58.08
MFCD00005167
AHHWIHXENZJRFG-UHFFFAOYSA-N
trimethylene oxide,1,3-propylene oxide,1,3-epoxypropane,oxacyclobutane,cyclooxabutane,trimethylenoxid,oxetan,1,3-trimethylene oxide,propane, 1,3-epoxy,trimethylenoxid german
10423
CHEBI:30965
oxetane
C1COC1
503-30-0
C3H6O
58.08
MFCD00005167
AHHWIHXENZJRFG-UHFFFAOYSA-N
trimethylene oxide,1,3-propylene oxide,1,3-epoxypropane,oxacyclobutane,cyclooxabutane,trimethylenoxid,oxetan,1,3-trimethylene oxide,propane, 1,3-epoxy,trimethylenoxid german
10423
CHEBI:30965
oxetane
C1COC1
Boiling Point 50°C
Specific Gravity 0.89
Flash Point <1°C
Density 0.8900g/mL
Refractive Index 1.3920 to 1.394
Packaging Glass bottle
Quantity 25mL
Infrared Spectrum Authentic
Color Colorless
Assay Percent Range 97%
Beilstein 17, 6
Fieser 01,1232
Merck Index 15, 9886
Formula Weight 58.08
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Trimethylene oxide, 97%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-964-3