Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethyl orthoformate, 99%, Acros Organics™

Click to view available options
Quantity:
100g
500g
1kg
5kg
Packaging:
Glass bottle
Plastic drum
149-73-5
C4H10O3
106.12
MFCD00008483
PYOKUURKVVELLB-UHFFFAOYSA-N
trimethyl orthoformate,methyl orthoformate,methane, trimethoxy,orthoformic acid, trimethyl ester,orthoformic acid methyl ester,orthomravencan methylnaty,methoxymethylal,methylester kyseliny orthomravenci,unii-xam28819yj,orthoformic acid trimethyl ester
9005
trimethoxymethane
COC(OC)OC
149-73-5
C4H10O3
106.12
MFCD00008483
PYOKUURKVVELLB-UHFFFAOYSA-N
trimethyl orthoformate,methyl orthoformate,methane, trimethoxy,orthoformic acid, trimethyl ester,orthoformic acid methyl ester,orthomravencan methylnaty,methoxymethylal,methylester kyseliny orthomravenci,unii-xam28819yj,orthoformic acid trimethyl ester
9005
trimethoxymethane
COC(OC)OC
Alkylation reagent
Refractive Index 1.3780 to 1.3800
Flash Point 13°C
Infrared Spectrum Authentic
Quantity 5kg
Melting Point -53.0°C
Density 0.9700g/mL
Color Colorless
Packaging Plastic drum
Boiling Point 101.0°C to 102.0°C
Specific Gravity 0.97
Assay Percent Range 99%
Linear Formula CH(OCH3)3
Beilstein 02, 19
Fieser 03,313; 04,540; 05,714; 08,507; 10,425; 14,330; 15,342
Merck Index 15, 6980
Solubility Information Solubility in water: 10g/L (hydrolysis). Other solubilities: soluble in organic solvents
Formula Weight 106.12
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Trimethyl orthoformate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger