Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylgermanium iodide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1066-38-2
C3H9GeI
244.639
MFCD00013700
KFLWSIOBXYVXBS-UHFFFAOYSA-N
trimethylgermanium iodide,trimethyliodogermane,iodo trimethyl germane
2734132
iodo(trimethyl)germane
C[Ge](C)(C)I
1066-38-2
C3H9GeI
244.639
MFCD00013700
KFLWSIOBXYVXBS-UHFFFAOYSA-N
trimethylgermanium iodide,trimethyliodogermane,iodo trimethyl germane
2734132
iodo(trimethyl)germane
C[Ge](C)(C)I
Quantity 5g
Packaging Glass bottle
Density 1.8150g/mL
Infrared Spectrum Authentic
Color Golden
Assay Percent Range 98%
Linear Formula (CH3)3GeI
Formula Weight 244.6
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material Trimethylgermanium iodide
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye damage.
May damage the unborn child.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger