Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Trimethylhydroquinone, 98%

Click to view available options
Quantity:
100g
500g
700-13-0
C9H12O2
152.193
MFCD00002346
AUFZRCJENRSRLY-UHFFFAOYSA-N
trimethylhydroquinone,2,3,5-trimethylhydroquinone,1,4-benzenediol, 2,3,5-trimethyl,pseudocumohydroquinone,psi-cumohydroquinone,2,3,6-trimethylhydroquinone,2,3,5-trimethyl-benzene-1,4-diol,hydroquinone, trimethyl,2,3,5-trimethyl-1,4-benzenediol,.psi.-cumohydroquinone
12785
2,3,5-trimethylbenzene-1,4-diol
CC1=CC(=C(C(=C1O)C)C)O
700-13-0
C9H12O2
152.193
MFCD00002346
AUFZRCJENRSRLY-UHFFFAOYSA-N
trimethylhydroquinone,2,3,5-trimethylhydroquinone,1,4-benzenediol, 2,3,5-trimethyl,pseudocumohydroquinone,psi-cumohydroquinone,2,3,6-trimethylhydroquinone,2,3,5-trimethyl-benzene-1,4-diol,hydroquinone, trimethyl,2,3,5-trimethyl-1,4-benzenediol,.psi.-cumohydroquinone
12785
2,3,5-trimethylbenzene-1,4-diol
CC1=CC(=C(C(=C1O)C)C)O
Applications
Trimethylhydroquinone is used for synthesizing vitamin E and used as medical intermediate.

Solubility
Soluble in water (2 g/L @ 20°C.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Flash Point 140°C (284°F)
Quantity 100g
Melting Point 169°C to 174°C
UN Number UN3077
Beilstein 1909183
Formula Weight 152.2
Percent Purity 98%
Chemical Name or Material Trimethylhydroquinone

GHS H Statement
H318-H332-H317-H335-H315
Causes serious eye damage.
Harmful if inhaled.
May cause an allergic skin reaction.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : MX7820000

TSCA : TSCA